Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse ErbB4/Her4 Fc Chimera Protein

Catalog No. 4387ER050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4387ER050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 4387ER050 Supplier R&D Systems™ Supplier No. 4387ER050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse ErbB4/Her4 Fc Chimera Protein is derived from NS0. The Recombinant Mouse ErbB4/Her4 Fc Chimera Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 13869
Molecular Weight (g/mol) 97.5 kDa (monomer)
Name ErbB4/Her4 Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse ErbB4/Her4 protein, (N-terminus) Mouse ErbB4 (Gln26-Val657) Accession &Num;Q61527 IEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 85%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.