Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Fas Ligand/TNFSF6 Protein

Catalog No. 526SA050
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Molecular Weight (g/mol):
18 kDa
18 kDa (monomer)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation Molecular Weight (g/mol)
526SA050 Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 18 kDa (monomer)
526SA050CF Lyophilized from a 0.2μm filtered solution in PBS. 18 kDa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 526SA050 Supplier R&D Systems™ Supplier No. 526SA050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse Fas Ligand/TNFSF6 Protein is derived from NS0. The Recombinant Mouse Fas Ligand/TNFSF6 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 14103
Molecular Weight (g/mol) 18 kDa (monomer)
Name Fas Ligand/TNFSF6
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse Fas Ligand/TNFSF6 protein Pro132-Leu279,with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.