Learn More
Description
Specifications
Specifications
| Accession Number | Q9JJN1 |
| Formulation | Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 and EDTA. |
| Gene ID (Entrez) | 56636 |
| Molecular Weight (g/mol) | 20kDa |
| Name | FGF-21 |
| Preparation Method | Reconstitute at 100μg/mL in PBS. |
| Quality Control Testing | SDS-PAGE: 22-25kDa, reducing conditions |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse FGF-21 protein Tyr30-Ser210 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
