Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse FGFR3 alpha (IIIb) Fc Chimera Protein

Catalog No. 9089FR050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-89F-R050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 90-89F-R050 Supplier R&D Systems™ Supplier No. 9089FR050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse FGF R3 alpha (IIIb) Fc Chimera Protein is derived from NS0. The Recombinant Mouse FGF R3 alpha (IIIb) Fc Chimera Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number NP_001156689
For Use With (Application) Bioactivity
Gene ID (Entrez) 2261
Molecular Weight (g/mol) Observed M.W.: 83-101kDa, reducing conditions, Theoretical M.W.: 63kDa
Name FGFR3
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse FGF R3 protein, (N-terminus) Mouse FGF R3 (IIIb) (Ala33-Tyr369) Accession &Num; NP_001156689 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias CD333, CEK, EC 2.7.10, FGF R3, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4, JTK4thanatophoric dwarfism
Common Name FGF R3
Gene Symbol Fgfr3
Biological Activity Measured by its ability to inhibit FGF acidic-dependent proliferation of BaF3 mouse pro-B cells transfected with human FGF R3 (IIIc). The ED50 for this effect is typically 0.2-1.2μg/mL.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Mouse FGF R3 (IIIb) (Ala33-Tyr369) Accession # NP_001156689, N-terminus; IEGRMD; Human IgG1 (Pro100-Lys330), C-terminus
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.