Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse FSH alpha/beta Protein

Catalog No. 8576FS010CF
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
85-76F-S010CF 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 85-76F-S010CF Supplier R&D Systems™ Supplier No. 8576FS010/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse FSH alpha/beta Protein is derived from CHO. The Recombinant Mouse FSH alpha/beta Protein has been validated for the following applications: Bioactivity.

Specifications

For Use With (Application) Bioactivity
Gene ID (Entrez) 2488
Molecular Weight (g/mol) Observed M.W.: 40-55kDa, reducing conditions, Theoretical M.W.: 24kDa
Quantity 10 μg
Source Chinese Hamster Ovary cell line,CHO-derived mouse FSH alpha/beta protein, (N-terminus) Mouse FSH beta (His20-Glu130) Accession &Num; Q60687 GGGSGGGSGGGS Mouse FSH alpha (Leu25-Ser120) Accession &Num; P01216 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Common Name FSH alpha/beta
Gene Symbol Fshb
Biological Activity Measured by its ability to induce cAMP accumulation in HEK293 human embryonic kidney cells transfected with human FSH R. The ED50 for this effect is typically 0.1-0.5ng/mL.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Mouse FSH beta (His20-Glu130) Accession # Q60687,N-terminus; GGGSGGGSGGGS; Mouse FSH alpha (Leu25-Ser120) Accession # P01216, C-terminus
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.