Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Galectin-2 Protein

Catalog No. 8579GA050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8579GA050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8579GA050 Supplier R&D Systems™ Supplier No. 8579GA050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Galectin-2 Protein is derived from HEK293. The Recombinant Mouse Galectin-2 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number AAH27504
For Use With (Application) Bioactivity
Gene ID (Entrez) 3957
Molecular Weight (g/mol) Observed M.W.: 12-14kDa, reducing conditions, Theoretical M.W.: 15kDa
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived mouse Galectin-2 protein Ser2-Glu130
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias Beta-galactoside-binding lectin L-14-II, Gal-2, GAL2, galectin 2, galectin-2, Galectin2, HL14gal-2, Lactose-binding lectin 2, lectin, galactoside-binding, soluble, 2, LGALS2, MGC75071, S-Lac lectin 2
Common Name Galectin-2
Gene Symbol Lgals2
Biological Activity Measured by the ability of the immobilized protein to support the adhesion of EOMA mouse endothelial cells. The ED50 for this effect is typically 0.5-2.5μg/mL. Measured by its ability to agglutinate human red blood cells. Hadari, Y.R. et al. (2000) J. Cell Sci. 113:2385. The ED50 for this effect is typically 2.5-12.5μg/mL.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Ser2-Glu130
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.