Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Galectin-3 Protein

Catalog No. 1197GA01M
Change view
Click to view available options
Quantity:
1 mg
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS, EDTA and DTT with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS, EDTA and DTT.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
11-97G-A01M 1 mg Lyophilized from a 0.2 μm filtered solution in PBS, EDTA and DTT with BSA as a carrier protein.
11-97G-A01MCF 1 mg Lyophilized from a 0.2 μm filtered solution in PBS, EDTA and DTT.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 11-97G-A01M Supplier R&D Systems™ Supplier No. 1197GA01M
Add to Cart
Add to Cart
This item is not returnable. View return policy

A New and Improved rm Galectin-3 is Now Available! It is expressed in mammalian cells with a lower endotoxin spec.

Measured by its ability to agglutinate human red blood cells. Hadari, Y.R. et al. (2000) J. Cell Sci. 113:2385. The ED50 for this effect is 5-20 μg/mL.

Specifications

Accession Number NP_034835
Characterization Bioactivity
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS, EDTA and DTT with BSA as a carrier protein.
Gene ID (Entrez) 3958
Molecular Weight (g/mol) M.W. (theoretical): 27.3 kDa
Quantity 1 mg
Source E. coli-derived mouse Galectin-3 protein Ala2-Ile264
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 3 months, 2°C to 8°C under sterile conditions after reconstitution.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias AGE-R3, Carbohydrate-binding protein 35, CBP 35,35kDa lectin, CBP35, Gal-3, GAL3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, Galectin3, GALIG, IgE-binding protein, L29, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, LGALS3, Mac-2, Mac-2 antigen, MAC2GAL3
Gene Symbol Lgals3
Product Type Recombinant Protein
Conjugate Unconjugated
Species Mouse
Reconstitution Reconstitute at 250 μg/mL in sterile PBS containing (at least 0.1% human or bovine serum albumin.)
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Galectin-3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.