Learn More
Abcam Recombinant Mouse Galectin 3 Protein, Active, 10 μg
Description
- Protein Type: Recombinant mouse Galectin 3 (Full Length, 1 to 264 aa)
- Expression System: Escherichia coli
- Purity: >95%
- Concentration: 0.5 mg/mL
- Tested Applications: FuncS, MS, SDS-PAGE
- Formulation: Liquid, pH 8
- Storage Buffer: 50% Glycerol, 0.87% Sodium chloride, 0.32% Tris HCl, 0.06% EDTA, 0.02% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Content And Storage | Store at -20°C. |
| Form | Liquid |
| Label or Dye | Unconjugated |
| Reactivity | Mouse |
| Shipping Condition | Blue Ice |
| Target Molecule | Galectin 3 |
| Concentration | 0.5 mg/mL |
| Formulation | 50% Glycerol, 0.87% Sodium chloride, 0.32% Tris HCl, 0.06% EDTA, 0.02% (R*,R*)-1,4-Dimercaptobutan-2,3-diol, pH 8 |
| Quantity | 10 μg |
| For Use With (Application) | SDS-PAGE, FuncS, Mass Spec |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.