Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Glypican 6 Fc Chimera Protein

Catalog No. 9429GP050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
94-29G-P050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 94-29G-P050 Supplier R&D Systems™ Supplier No. 9429GP050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Glypican 6 Fc Chimera Protein is derived from NS0. The Recombinant Mouse Glypican 6 Fc Chimera Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q9R087
Characterization Bioactivity
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 10082
Molecular Weight (g/mol) 83kDa (Asp24-Val513/mFc), 44kDa (Ser355-Val513/mFc) and 38kDa (Asp24-Arg354)
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse Glypican 6 protein, (N-terminus) Mouse Glypican 6 (Asp24-Val513) Accession &Num; Q9R087 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10EU per 1μg of the protein by the LAL method.
Gene Alias glypican 6, glypican proteoglycan 6, glypican-6, GPC6,mgC126288, OMIMD1
Product Type Recombinant Protein
Conjugate Unconjugated
Species Mouse
Reconstitution Reconstitute at 500μg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.