Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse GM-CSF Protein

Catalog No. 415ML01MCF
Change view
Click to view available options
Quantity:
1 mg
20 μg
5 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-5ML0-1MCF 1 mg
41-5ML0-05CF 5 μg
41-5ML0-20CF 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 41-5ML0-1MCF Supplier R&D Systems™ Supplier No. 415ML01M/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse GM-CSF Protein is derived from E. coli. The Recombinant Mouse GM-CSF Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q14AD9
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS. *1 mg pack size (01M) is supplied as a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 1437
Molecular Weight (g/mol) 14.3 kDa
Quantity 1 mg
Source <EM>E. coli</EM>-derived mouse GM-CSF protein Ala18-Lys141, with an N-terminal Met
Storage Requirements Manual defrost freezer; Avoid repeated freeze-thaw cycles; 12 mo. from receipt, -20 to -70°C as supplied; 1 mo, 2 to 8°C under sterile conditions after reconstitution; 3 mo, -20 to -70°C under sterile conditions after reconstitution
Regulatory Status RUO
Endotoxin Concentration <1.0 EU per 1 μg of the protein by the LAL method.
Gene Alias colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim
Gene Symbol Csf2
Conjugate Unconjugated
Cross Reactivity GM-CSF
Species Mouse
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Buffer Lyophilized from a 0.2μm filtered solution in PBS.
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.