Learn More
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl and CaCl 2 . |
| Gene ID (Entrez) | 54426 |
| Molecular Weight (g/mol) | 32 kDa & 28 kDa |
| Name | HGF Activator |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse HGF Activator protein Met1-Ser653 with a C-terminal 10-His tag The pro form was purified,activated and further purified. |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Structural Form | Active form |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
