Learn More
R&D Systems™ Recombinant Mouse HGFR/c-MET Fc Chimera (NS0-expressed)
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 17295 |
| Molecular Weight (g/mol) | 96 & 32 kDa (monomer) |
| Name | HGFR/c-MET |
| Quantity | 100 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse HGF R/c-MET protein, (N-terminus) Mouse HGF R/c-Met (Met1-Asn929) Accession &Num; P16056 DIEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | AUTS9, c-Met, cMET, EC 2.7.10, EC 2.7.10.1, hepatocyte growth factor receptor, HGF R, HGF receptor, HGF/SF receptor, HGFR, MET, Met (c-Met), met proto-oncogene (hepatocyte growth factor receptor), met proto-oncogene tyrosine kinase, oncogene MET, Proto-oncogene c-Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.