Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse IDO Protein
SDP

Catalog No. 9157AO010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-57A-O010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 91-57A-O010 Supplier R&D Systems™ Supplier No. 9157AO010
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Indoleamine 2,3-dioxygenase

Measured by its ability to oxidize L-tryptophan to N-formyl-kynurenine. The specific activity is >1,000 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number P28776
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Sodium Acetate, NaCl and Glycerol.
Gene ID (Entrez) 3620
Molecular Weight (g/mol) M.W. (Observed): 41 kDa, reducing conditions; M.W. (theoretical): 46 kDa
Quantity 10 μg
Source E. coli-derived mouse Indoleamine 2,3-dioxygenase/IDO protein Ala2-Pro407, with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias 3dioxygenase, EC 1.13.11.52, IDO1, IDOIDO-1, INDO, INDOindole 2,3-dioxygenase, Indoleamine 2, indoleamine 2,3-dioxygenase 1, indoleamine-pyrrole 2,3 dioxygenase, Indoleamine-pyrrole 2,3-dioxygenase
Common Name Indoleamine 2,3-dioxygenase/IDO
Biological Activity Measured by its ability to oxidize L-tryptophan to N-formyl-kynurenine. The specific activity is >1,000 pmol/min/μg, as measured under the described conditions. See Activity Assay Protocol on RnDSystems.com.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System E. coli-derived Ala2-Pro407, with an N-terminal Met and 6-His tag
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Indoleamine 2,3-dioxygenase/IDO
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.