Learn More
Description
Specifications
Specifications
| Accession Number | K00020 |
| For Use With (Application) | Bioactivity |
| Formulation | 20mM Sodium Phosphate, pH 6.4; 300mM NaCl; 5% Glycerol |
| Molecular Weight (g/mol) | Predicted MW: 19.6kDa |
| Source | Gene was obtained from mouse DNA expressed in mammalian cells. Accession # K00020 |
| Storage Requirements | Store the unopened product at -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Gene Alias | Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956 |
| Conjugate | Unconjugated |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
