Learn More
R&D Systems™ Recombinant Mouse IFN-beta Protein
SDP
Catalog No. 124001 Shop All R&D Systems Products
This item is not returnable. View return policy
Catalog No. 124001 Supplier R&D Systems™ Supplier No. 124001
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Specifications
Storage Requirements Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Gene Alias Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956