Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse IFN-beta Protein
SDP

Catalog No. 8234MB010CF Shop All R&D Systems Products
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS and Tween™ 80
Lyophilized from a 0.2μm filtered solution in PBS and Tween™ 80 with BSA as a carrier protein
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
82-34M-B010CF Lyophilized from a 0.2μm filtered solution in PBS and Tween™ 80
82-34M-B010 Lyophilized from a 0.2μm filtered solution in PBS and Tween™ 80 with BSA as a carrier protein
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 82-34M-B010CF Supplier R&D Systems™ Supplier No. 8234MB010/CF
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse IFN-beta Protein is derived from HEK293. The Recombinant Mouse IFN-beta Protein has been validated for the following applications: Bioactivity.

Order Info

The product is shipped at ambient temperature, upon receipt, store immediately

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS and Tween™ 80
Quantity 10 μg
Source Human embryonic kidney cell,HEK293-derived mouse IFN-beta protein Ile22-Asn182
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956
Conjugate Unconjugated
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.