Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse IFN-kappa Protein

Catalog No. 8437MK010CF
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in Sodium Acetate, NaCl and EDTA with BSA as a carrier protein
Lyophilized from a 0.2μm filtered solution in Sodium Acetate, NaCl and EDTA.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
84-37M-K010CF Lyophilized from a 0.2μm filtered solution in Sodium Acetate, NaCl and EDTA.
84-37M-K010 Lyophilized from a 0.2μm filtered solution in Sodium Acetate, NaCl and EDTA with BSA as a carrier protein
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 84-37M-K010CF Supplier R&D Systems™ Supplier No. 8437MK010/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse IFN-kappa Protein is derived from NS0. The Recombinant Mouse IFN-kappa Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q7TSL0
Formulation Lyophilized from a 0.2μm filtered solution in Sodium Acetate, NaCl and EDTA.
Gene ID (Entrez) 56832
Molecular Weight (g/mol) 21kDa
Name IFN-kappa
Preparation Method Reconstitute at 200μg/mL in sterile water.
Quality Control Testing SDS-PAGE: 19-22kDa, reducing conditions
Quantity 10 μg
Source Mouse myeloma cell line,NS0-derived mouse IFN-kappa protein Leu22-Lys199
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Leu22
Without Additives Carrier Protein
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE with silver staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.