Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse IFNAA Protein

Catalog No. 10150IF050
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-150-IF050 50 μg
10-150-IF010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 10-150-IF050 Supplier R&D Systems™ Supplier No. 10150IF050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured in an anti-viral assay using L-929 mouse fibroblast cells infected with encephalomyocarditis (EMC) virus.

  • The ED50 for this effect is <15 pg/mL.
  • Storage:
    • Use a manual defrost freezer and avoid repeated freeze-thaw cycles
    • 12 months from date of receipt, -20 to -70°C as supplied
    • 1 month, 2 to 8°C under sterile conditions after reconstitution
    • 3 months, -20 to -70°C under sterile conditions after reconstitution

Specifications

Accession Number Q61718
For Use With (Application) Bioactivity
Gene ID (Entrez) 242517
Molecular Weight (g/mol) 16-21 kDa, under reducing conditions
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived mouse IFNAA protein Cys24-Glu190
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles
Endotoxin Concentration < 0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Species Mouse
Reconstitution Reconstitute at 100 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose
Purity or Quality Grade > 95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.