Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse IFNAB Protein

Catalog No. 10151IF050
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-151-IF050 50 μg
10-151-IF010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 10-151-IF050 Supplier R&D Systems™ Supplier No. 10151IF050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human embryonic kidney cell, HEK293-derived mouse IFNAB protein

  • The interferons (IFN) are a family of cytokines with potent antiviral, antiproliferative and immunomodulatory properties, and are classified based on their binding specificity to cell surface receptors
  • The mouse IFNAB consists of 190 amino acids (aa) including a 23 aa signal peptide and a 167 aa IFNAB main chain.
  • It was found that IFNAB plays an important role in the regulation of DNA synthesis in colong-stimulating factor (CSF)-1-stimulated macrophage, as well as mediates the inhibitory efffects of LPS and TNF alpha

Specifications

Accession Number AAA58254.1
For Use With (Application) Bioactivity
Gene ID (Entrez) 15974
Molecular Weight (g/mol) 19 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived mouse IFNAB protein Cys24-Glu190
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after opening. 3 months, -20 to -70°C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 100 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.