Learn More
R&D Systems™ Recombinant Mouse IGSF4C/SynCAM4 Fc Chimera Protein
Shop All Novus Biologicals Products

Description
- Derived from NS0
- Validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | Q8R464 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 199731 |
| Molecular Weight (g/mol) | 81 to 93 kDa, reducing conditions |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived mouse IGSF4C/SynCAM4 protein. Mouse IGSF4C (Gln25-Ala324) Accession # Q8R464IEGRMDHuman IgG1 (Pro100-Lys330)N-terminus C-terminus |
| Storage Requirements | 12 months from date of receipt, -20 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution 3 months, -20 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | < 0.10 EU per 1 μg of the protein by the LAL method |
| Conjugate | Unconjugated |
| Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.