Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse IGSF9 Protein

Catalog No. 9140S9050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-40S-9050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 91-40S-9050 Supplier R&D Systems™ Supplier No. 9140S9050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse IGSF9 Protein is derived from NS0. The Recombinant Mouse IGSF9 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q05BQ1
For Use With (Application) Bioactivity
Gene ID (Entrez) 57549
Molecular Weight (g/mol) Observed M.W.: 95-110kDa, reducing conditions, Theoretical M.W.: 78kDa
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse IGSF9 protein Arg21-Leu730,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias Dasm1, FP18798, IGSF9A, immunoglobulin superfamily member 9A, immunoglobulin superfamily, member 9, KIAA1355, Nrt1, Protein Turtle Homolog A, TUTLA
Common Name IGSF9
Biological Activity Measured by its ability to inhibit neurite outgrowth of dissociated E13 chick embryonic dorsal root ganglia (DRG) neurons. Able to significantly inhibit neurite outgrowth when immobilized as a 3μL droplet containing 150 ng on a nitrocellulose-coated microplate
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Arg21-Leu730, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.