Learn More
R&D Systems™ Recombinant Mouse IL-11 Protein
Description
Specifications
Specifications
| Formulation | PBS, pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl), 0.1% BSA |
| Gene ID (Entrez) | 16156 |
| Molecular Weight (g/mol) | 19 kDa |
| Name | IL-11 |
| Quantity | 25 μg |
| Source | E. coli-derived mouse IL-11 protein Gly23-Leu199,with an N-terminal Met and Pro25-Leu199 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | Adipogenesis inhibitory factor, AGIF, AGIFoprelvekin, IL11, IL-11Oprelvekin, interleukin 11, interleukin-11, Oprelvekin |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.