Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse IL-12 (linked heterodimer) Protein

Catalog No. 1051ML010CF
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-51M-L010CF 10 μg
10-51M-L050CF 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 10-51M-L010CF Supplier R&D Systems™ Supplier No. 10051ML010/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

GS Linker, Defined heterodimer stoichiometry.

Measured in a cell proliferation assay using PHA-activated mouse splenocytes. Mattner, F. et al. (1993) Eur. J. Immunol. 23:2202. The ED50 for this effect is 0.01-0.1 ng/mL.

Specifications

Accession Number P43432, P43431
For Use With (Application) Bioactivity
Gene ID (Entrez) 3592
Molecular Weight (g/mol) 58 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 10 μg
Source

Mouse myeloma cell line, NS0-derived mouse IL-12 protein

Human IL-12 p40 (Met23-Ser335) Accession # P43432GGGGSGGGGSGGGGSMouse IL-12 p35 (Arg23-Ala215) Accession # P43431N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, ≤ -20°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 100 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.