Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse IL-15 Protein

Catalog No. 447ML010
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS and Tween™ 20.
Lyophilized from a 0.2μm filtered solution in Tris and NaCl with BSA as a carrier protein.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
447ML010 Lyophilized from a 0.2μm filtered solution in Tris and NaCl with BSA as a carrier protein.
447ML010CF Lyophilized from a 0.2μm filtered solution in PBS and Tween™ 20.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 447ML010 Supplier R&D Systems™ Supplier No. 447ML010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse IL-15 Protein is derived from E. coli. The Recombinant Mouse IL-15 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Tris and NaCl with BSA as a carrier protein.
Gene ID (Entrez) 16168
Molecular Weight (g/mol) 13.4 kDa
Name IL-15
Quantity 10 μg
Source E. coli-derived mouse IL-15 protein Asn49-Ser162,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.