Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse IL-15 Protein
SDP

Catalog No. p-7183008 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.1 mg
0.5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002953 0.05 mg
NB002955 0.5 mg
NB002954 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB002953 Supplier Novus Biologicals™ Supplier No. NBP2774970.05MG

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

Mouse IL-15 stimulates cell proliferation of the CTLL2 mouse cytotoxic T cells. The ED50 range is less or equal to 20 ng/ml.{ATGP3883-BA.JPG}

Specifications

Concentration 0.5 mg/mL
For Use With (Application) SDS-PAGE
Formulation PBS (pH7.4), 10% glycerol
Gene ID (Entrez) 3600
Molecular Weight (g/mol) 17.6 kDa
Quantity 0.05 mg
Source E. coli
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene Alias IL-15MGC9721, interleukin 15, interleukin-15
Gene Symbol IL15
Buffer PBS (pH7.4), 10% glycerol
Endotoxin Level < 1.0 EU per 1 μg of protein (determined by LAL method)
Purity or Quality Grade >90% SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.