Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse IL-18/IL-1F4 Protein

Catalog No. 9139IL050
Change view
Click to view available options
Quantity:
10 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-39I-L050 50 μg
91-39I-L010 10 μg
91-39I-L500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 91-39I-L050 Supplier R&D Systems™ Supplier No. 9139IL050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse IL-18/IL-1F4 Protein is derived from E. coli. The Recombinant Mouse IL-18/IL-1F4 Protein has been validated for the following applications: Bioactivity.

Order Info

The product is shipped at ambient temperature.

Specifications

Accession Number P70380
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS, EDTA and DTT with BSA as a carrier protein.
Gene ID (Entrez) 3606
Molecular Weight (g/mol) 18 kDa
Name Iboctadekin, IFN-gamma-inducing factor, IGIF, IGIFIL-1 gamma, IL18, IL-18MGC12320, IL-1F4, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18
Quantity 50 μg
Source E. coli-derived mouse IL-18/IL-1F4 protein Asn36-Ser192
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of protein by LAL method
Biological Activity Measured by its ability to induce mouse IFN-gamma secretion by activated mouse T cells. The ED50 for this effect is typically 0.06-0.36 ng/mL.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.