Learn More
R&D Systems™ Recombinant Mouse IL-18/IL-1F4 Protein
Description
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized in PBS containing 1% sucrose. |
| Molecular Weight (g/mol) | Predicted MW: 18kDa |
| Quantity | 25 μg |
| Source | E. coli-derived mouse IL-18/IL-1F4 protein 36-192 aa |
| Storage Requirements | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Reconstitution | Reconstitute in 250μL of ice-cold distilled water on ice. |
| Purity or Quality Grade | >90% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.