Learn More
R&D Systems™ Recombinant Mouse IL-1ra/IL-1F3 Protein

Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Sodium Citrate, NaCl, EDTA and DTT with BSA as a carrier protein. |
| Gene ID (Entrez) | 16181 |
| Molecular Weight (g/mol) | 17 kDa |
| Name | IL-1ra/IL-1F3 |
| Quantity | 10 μg |
| Source | E. coli-derived mouse IL-1ra/IL-1F3 protein Arg8-Gln159,with an N-terminal Met |
| Storage Requirements | 12 months from date of receipt, ≤ -20° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, ≤ -20° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | >97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.