Learn More
R&D Systems™ Recombinant Mouse IL-2 Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Sodium Citrate and Tween™-80. |
| Gene ID (Entrez) | 16183 |
| Molecular Weight (g/mol) | 17.2 kDa |
| Name | IL-2 |
| Quantity | 500 μg |
| Source | E. coli-derived mouse IL-2 protein Ala21-Gln169,with and without an N-terminal Met |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Reconstitution | Reconstitute at 100-200μg/mL in sterile deionized water. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.