Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse IL-21 Protein

Catalog No. 594ML025CF
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-4ML0-25CF 25 μg
59-4ML1-00CF 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 59-4ML0-25CF Supplier R&D Systems™ Supplier No. 594ML025/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse IL-21 Protein is derived from E. coli. The Recombinant Mouse IL-21 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q9ES17.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS. *1 mg pack size (01M) is supplied as a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 59067
Molecular Weight (g/mol) 14.4 kDa
Quantity 25 μg
Source <EM>E. coli</EM>-derived mouse IL-21 protein Pro25-Ser146, with and without an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles; 12 months from date of receipt, -20 to -70°C as supplied; 3 months, 2 to 8°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.01 EU per 1 μg of the protein by the LAL method.
Gene Symbol Il21
Conjugate Unconjugated
Cross Reactivity IL-21
Species Mouse
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.