Learn More
Description
Specifications
Specifications
| Accession Number | P43432.1 (p40) & Q9EQ14.1 (p19) |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Molecular Weight (g/mol) | MolecularWeight-observed: 60-75 kDa, under reducing conditions., MolecularWeight-theroretical: 57 kDa |
| Quantity | 10 μg |
| Source | Human embryonic kidney cell, HEK293-derived mouse IL-23 protein Mouse IL-12p40 (Met22-Ser335) Accession # P43432.1 GSGSSRGGSGSGGSGGGGSKL Mouse IL-23p19 (Leu20-Ala196) Accession # Q9EQ14.1 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Reconstitution | Reconstitute at 100-500 μg/mL in PBS. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
