Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse IL-27 p28/IL-30 Protein
SDP

Catalog No. 7430ML010CF Shop All Novus Products
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in HEPES, NaCl and CHAPS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in HEPES, NaCl and CHAPS.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
7430ML010CF Lyophilized from a 0.2μm filtered solution in HEPES, NaCl and CHAPS.
7430ML010 Lyophilized from a 0.2μm filtered solution in HEPES, NaCl and CHAPS with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 7430ML010CF Supplier R&D Systems™ Supplier No. 7430ML010/CF

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse IL-27 p28/IL-30 Protein is derived from NS0. The Recombinant Mouse IL-27 p28/IL-30 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in HEPES, NaCl and CHAPS.
Gene ID (Entrez) 246779
Molecular Weight (g/mol) Observed Molecular Wt: 25-35kDa, reducing conditions, Predicted Molecular Wt.: 24.9kDa
Name IL-27 p28/IL-30
Quantity 10 μg
Source Mouse myeloma cell line,NS0-derived mouse IL-27 p28/IL-30 protein Phe29-Ser234,with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Product Line R&D Systems
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.