Learn More
R&D Systems™ Recombinant Mouse IL-3 Protein

Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 16187 |
| Molecular Weight (g/mol) | 15 kDa |
| Name | IL-3 |
| Quantity | 500 μg |
| Source | E. coli-derived mouse IL-3 protein Asp33-Cys166,with an N-terminal Met |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | Hematopoietic growth factor, IL3, IL-3MGC79398, interleukin 3 (colony-stimulating factor, multiple), interleukin-3, Mast cell growth factor, mast-cell growth factor, MCGF, MCGFMGC79399, MULTI-CSF, multilineage-colony-stimulating factor, Multipotential colony-stimulating factor, P-cell stimulating factor, P-cell-stimulating factor |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.