Learn More
Abcam Recombinant mouse IL-6 protein, 500UG
Supplier:  Abcam AB198572500UG
Encompass_Preferred
Recombinant mouse IL-6 protein is a Mouse Full Length protein, in the 25 to 211 aa range, expressed in Escherichia coli, with >95% purity, <= 1 EU/µg endotoxin level and suitable for SDS-PAGE, FuncS, HPLC.
Protein/ Peptide/ Enzyme Synonyms: Il-6, Il6, Interleukin-6, IL-6, B-cell hybridoma growth factor, Interleukin HP-1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-9370
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.