Learn More
Description
Specifications
Specifications
| Accession Number | Q80V26 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol. |
| Gene ID (Entrez) | 54928 |
| Molecular Weight (g/mol) | M.W. (Observed): 35-40 kDa, reducing conditions; M.W. (theoretical): 34 kDa |
| Name | Inositol Monophosphatase 3/IMPAD1 |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived mouse Inositol Monophosphatase 3/IMPAD1 protein Glu51-His356 with an N-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
