Learn More
R&D Systems™ Recombinant Mouse Integrin alpha 3 beta 1/VLA-3 Protein
Description
Includes
For Research Use Only
Order Info
Ships at ambient temperature.
Specifications
Specifications
| Accession Number | Q62470-1 (Integrin alpha 3) & P09055-1 (Integrin beta 1) |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS |
| Gene ID (Entrez) | 3675 |
| Molecular Weight (g/mol) | 116 kDa (Integrin alpha 3), 86 kDa (Integrin beta 1) |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived mouse Integrin alpha 3 beta 1/VLA-3 protein, (N-terminus) Mouse Integrin alpha 3 (Phe33-Glu993) Accession &Num; Q62470-1 His-Pro GGGSGGGS Acidic Tail 6-His tag Mouse Integrin beta 1 (Gln21-Asp728) Accession &Num; P09055-1 His-Pro GGGSGGGS Basic Tail (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.