Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Integrin alpha E beta 7 Protein

Catalog No. 8356A3050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
83-56A-3050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 83-56A-3050 Supplier R&D Systems™ Supplier No. 8356A3050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Integrin alpha E beta 7 Protein is derived from CHO. The Recombinant Mouse Integrin alpha E beta 7 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number ABD49099 (alpha E), P26011 (beta 7)
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Molecular Weight (g/mol) 130kDa (αE), 84kDa (β7)
Name Integrin alpha E beta 7
Preparation Method Reconstitute at 500μg/mL in sterile PBS.
Quality Control Testing SDS-PAGE: 112-128kDa and 142-176kDa, reducing conditions
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived mouse Integrin alpha E beta 7 protein, (N-terminus) Mouse Integrin alpha E (Phe20-Arg1113,Ser453Gly) Accession &Num; ABD49099 His-Pro GGGSGGGS Acidic Tail 6-His tag Mouse Integrin ss7 (Glu20-Arg724) Accession &Num; P26011 GGGSGGGS Basic Tail (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Phe20 & Thr183 (αE), Glu20 (β7)
Without Additives Carrier Protein
Structural Form Noncovalent heterodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE with silver staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.