Learn More
R&D Systems™ Recombinant Mouse Integrin alpha V beta 1 Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Molecular Weight (g/mol) | 115 kDa ( alphaV subunit) & 86.4 kDa (beta1 subunit) |
| Name | Integrin alpha V beta 1 |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived mouse Integrin alpha V beta 1 protein, (N-terminus) Mouse Integrin alpha V (Phe31-Val988) Accession &Num; P43406 HP GS Linker Acidic Tail HHHHHH Mouse Integrin beta 1 (Gln21-Asn728) Accession &Num; P09055 His GS Linker Basic Tail (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Noncovalently-linked heterodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.