Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Integrin alpha V beta 8 Protein

Catalog No. 8314AV050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
83-14A-V050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 83-14A-V050 Supplier R&D Systems™ Supplier No. 8314AV050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. Immobilized Recombinant Mouse Integrin alpha V beta 8 at 2 μg/mL can bind Recombinant Human LAP (TGF-beta 1) (Catalog # 246-LP ) with an apparent Kd <0.1 nM.

Specifications

Accession Number P43406 & Q0VBD0
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS and Trehalose.
Gene ID (Entrez) 3685
Molecular Weight (g/mol) M.W. (Observed): 95 - 170 kDa, reducing conditions; M.W. (theoretical): 115 kDa (alpha V subunit), 78 kDa (beta 8 subunit)
Name Integrin alpha V beta 8
Preparation Method Reconstitute at 500μg/mL in sterile PBS.
Quality Control Testing SDS-PAGE: 95 - 170kDa, reducing conditions
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived mouse Integrin alpha V beta 8 protein Mouse Integrin aV (Phe31-Val988) (N-terminus) Accession # P43406 His-Pro GGGSGGGS Acidic Tail 6-His tag Mouse Integrin ss8 (Glu43-Ser679) (N-terminus) Accession # Q0VBD0 His-Pro GGGSGGGS Basic Tail (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Phe31 (αV subunit) & Glu43 (β8 subunit)
Without Additives Carrier Protein
Structural Form Noncovalent heterodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL in sterile PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Integrin alpha V beta 8
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.