Learn More
Abcam Recombinant Mouse Interferon Beta, Full Length, 10 μg
Description
- Protein Type: Recombinant Mouse Interferon beta
- Host System: HEK 293 cells
- Sequence Length: Full length (30 to 379 aa)
- Applications: SDS-PAGE, HPLC, MS
- Purity: ≥95%
- Endotoxin Level: ≤0.005 EU/μg
- Form: Lyophilized; pH 7.4; contains 10.26% Trehalose, 0.727% Dibasic monohydrogen potassium phosphate, and 0.428% Potassium phosphate monobasic
- Animal-Free: Yes
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | Interferon Beta |
| Reactivity | Mouse |
| Label or Dye | Unconjugated |
| Buffer | 10.26% trehalose, 0.727% dibasic monohydrogen potassium phosphate, 0.428% potassium phosphate monobasic, pH7.4 |
| Content And Storage | Store at ambient temperature. |
| Form | Lyophilized |
| Formulation | Lyophilized from pH 7.4 buffer containing 10.26% Trehalose, 0.727% Dibasic monohydrogen potassium phosphate, and 0.428% Potassium phosphate monobasic |
| Quantity | 10 μg |
| For Use With (Application) | SDS-PAGE, HPLC, Mass Spec |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.