Learn More
Abcam Recombinant mouse Jagged1 protein (Fc Chimera Active), 50UG
Supplier:  Abcam AB10934650UG
Encompass_Preferred
Recombinant mouse Jagged1 protein (Fc Chimera Active) is a Mouse Fragment protein, in the 1 to 1067 aa range, expressed in HEK 293, with >90% purity, < 0.01 EU/µg endotoxin level and suitable for SDS-PAGE, Inhib, FuncS.
Protein/ Peptide/ Enzyme Synonyms: CD339, Protein jagged-1, Jagged1, Jag1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-5154
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.