Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse Kallikrein 8/Neuropsin Protein
SDP

Catalog No. NBP259574Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
250 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259574Y 250 μg
NBP25957450 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP259574Y Supplier Novus Biologicals™ Supplier No. NBP259574250UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids29-260 of Mouse Kallikrein 8/Neuropsin The Recombinant Mouse Kallikrein 8/Neuropsin Protein is derived from Baculovirus. The Recombinant Mouse Kallikrein 8/Neuropsin Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. PBS (pH 7.4) containing 10% glycerol
Gene ID (Entrez) 11202
Molecular Weight (g/mol) 26.5 kDa
Name Recombinant Mouse Kallikrein 8/Neuropsin Protein
Purification Method >90%, by SDS-PAGE
Quantity 250 μg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU per 1μg of protein (determined by LAL method)
Gene Alias EC 3.4.21, EC 3.4.21.118, hK8, HNP, kallikrein 8 (neuropsin/ovasin), kallikrein-related peptidase 8, KLK8 protein type 1, KLK8 protein type 2, Neuropsin, neuropsin type 1, neuropsin type 2, NP, NRPN, Ovasin, PRSS19kallikrein-8, Serine protease 19, Serine
Gene Symbol KLK8
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Mouse
Recombinant Recombinant
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.