Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Lefty-1 Protein

Catalog No. 994LF025
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
994LF025 Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
994LF025CF Lyophilized from a 0.2μm filtered solution in PBS.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 994LF025 Supplier R&D Systems™ Supplier No. 994LF025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Lefty-1 Protein is derived from NS0. The Recombinant Mouse Lefty-1 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 10637
Molecular Weight (g/mol) 26.9 kDa
Name Lefty-1
Quantity 25 μg
Source Mouse myeloma cell line,NS0-derived mouse Lefty-1 protein Mouse Lefty-1 Proregion (Leu22-Arg135) Accession &Num; NP_034224 6-His tag Mouse Lefty mature (Leu136-Pro368) Accession &Num; NP_034224 6-His tag Mouse Lefty mature (Leu136-Pro368) Accession &Num; NP_034224 N-terminus C-terminus A short DNA sequence encoding 6 histidine residues was inserted into the DNA sequence encoding mouse Lefty-1 between Arg135 and Leu136. Two peptides corresponding to mouse Lefty-1 propeptide and mature mouse Lefty-1 are present in the recombinant preparation.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.1 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.