Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse LIF Protein

Catalog No. 8878LF100CF
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8878LF100CF 100 μg
8878LF025CF 25 μg
8878LF500CF 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 8878LF100CF Supplier R&D Systems™ Supplier No. 8878LF100/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse LIF Protein is derived from E. coli. The Recombinant Mouse LIF Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P09056
For Use With (Application) Bioactivity
Gene ID (Entrez) 3976
Molecular Weight (g/mol) Observed M.W.: 20kDa, reducing conditions, Theoretical M.W.: 20kDa
Quantity 100 μg
Source E. coli-derived mouse LIF protein Pro25-Phe203
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDA, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI
Common Name LIF
Gene Symbol Lif
Biological Activity Measured by its ability to induce IL-6 secretion by M1 mouse myeloid leukemia cells. The ED50 for this effect is typically 0.1-0.6ng/mL.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System E. coli-derived Pro25-Phe203
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.