Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse LIGHT/TNFSF14 Protein

Catalog No. 1794LT025CF
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
1794LT025CF Lyophilized from a 0.2 μm filtered solution in PBS.
1794LT025 Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 1794LT025CF Supplier R&D Systems™ Supplier No. 1794LT025/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cytotoxicity assay using HT-29 human colon adenocarcinoma cells Yu, K.Y. et al. (1999) J. Biol. Chem. 274:13733; Harrop, J.A. et al. (1998) J. Biol. Chem. 273:27548; Zhai, Y. et al. (1998) J. Clin. Invest. 102:1142. The ED50 for this effect is 2-5 μg/mL in the presence of a cross-linking antibody, Mouse Anti-polyHistidine Monoclonal Antibody (Catalog # MAB050 ) and recombinant human IFN-gamma .

Specifications

Accession Number Q9QYH9
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 8740
Molecular Weight (g/mol) M.W. (Observed): 24-34 kDa, reducing conditions; M.W. (theoretical): 21 kDa
Name LIGHT/TNFSF14
Quantity 25 μg
Source Mouse myeloma cell line, NS0-derived mouse LIGHT/TNFSF14 protein MHHHHHHHHHH GGGSGGGSGGGS IEGR Mouse LIGHT (Asp72-Val239) (N-terminus) Accession # Q9QYH9 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol Tnfsf14
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein LIGHT/TNFSF14
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.