Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Lymphotoxin alpha2/beta1 Protein

Catalog No. 1008LY010CF
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
1008LY010CF Lyophilized from a 0.2μm filtered solution in PBS.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 1008LY010CF Supplier R&D Systems™ Supplier No. 1008LY010/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse Lymphotoxin alpha2/beta1 Protein is derived from Sf 21 (baculovirus). The Recombinant Mouse Lymphotoxin alpha2/beta1 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 16992
Molecular Weight (g/mol) 18.5 kDa (LT alpha), 18.4 kDa (LTbeta)
Name Lymphotoxin α2/β1
Quantity 10 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse Lymphotoxin protein Leu34-Leu202 (LT alpha ) & Gly136-Gly306 (LT beta )
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Noncovalently-linked heterotrimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.