Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Lysosomal alpha-Glucosidase/GAA, CF

Catalog No. 11400GH025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-400-GH025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-400-GH025 Supplier R&D Systems™ Supplier No. 11400GH025
Add to Cart
Add to Cart
This item is not returnable. View return policy

His-tag

Measured by its ability to release glucose from starch. The specific activity is >5000 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number P70699.2
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 2548
Molecular Weight (g/mol) M.W. (Observed): 97-107 kDa, under reducing conditions, M.W. (Theroretical): 99 kDa
Quantity 25 μg
Source Chinese Hamster Ovary cell line, CHO-derived mouse Lysosomal alpha-Glucosidase protein Glu70-Ser953 with a N-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias Acid maltase, Aglucosidase alfa, EC 3.2.1.20, glucosidase, alpha; acid, LYAG, lysosomal alpha-glucosidase
Conjugate Unconjugated
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein LYAG/GAA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.