Learn More
Abcam Recombinant mouse M-CSF protein, 100UG
Supplier:  Abcam AB129146100UG
Encompass_Preferred
Recombinant mouse M-CSF protein is a Mouse Fragment protein, in the 33 to 187 aa range, expressed in Escherichia coli, with >=95% purity, <= 1 EU/µg endotoxin level and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: Csfm, Csf1, Macrophage colony-stimulating factor 1, CSF-1, MCSF, Proteoglycan macrophage colony-stimulating factor, PG-M-CSF
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4162
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.