Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Mesothelin (aa 298-600) His Protein

Catalog No. 8604MS050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
86-04M-S050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 86-04M-S050 Supplier R&D Systems™ Supplier No. 8604MS050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Mesothelin Protein is derived from CHO. The Recombinant Mouse Mesothelin Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q61468
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS with trehalose.
Gene ID (Entrez) 10232
Molecular Weight (g/mol) Observed M.W.: 38-55kDa, reducing conditions, Theoretical M.W.: 35kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived mouse Mesothelin protein Asp298-Ser600,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Termination C-Terminal
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias CAK1, CAK1 antigen, megakaryocyte potentiating factor, mesothelin, MPF, MPFSMRP, MSLN, Pre-pro-megakaryocyte-potentiating factor, SMR, soluble MPF mesothelin related protein
Common Name Mesothelin
Gene Symbol Msln
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Mouse Mesothelin is immobilized at 2μg/mL (100μL/well), the concentration of Recombinant Human CA125/MUC16 that produces 50% optimal binding response is approximately 0.4-2.4ng/mL.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Activity Measured by its binding ability in a functional ELISA. When Recombinant Mouse Mesothelin C-Terminal (aa 298-600) His-tag (Catalog 8604-MS) is immobilized at 2 μg/mL (100 μL/well), the concentration of Recombinant Human CA125/MUC16 (Catalog # 5609-MU ) that produces 50% optimal binding response is approximately 0.4-2.4 ng/mL.
Expression System Chinese Hamster Ovary cell line, CHO-derived Asp298-Ser600, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE with silver staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.