Learn More
R&D Systems™ Recombinant Mouse MuSK Fc Chimera Protein
Description
Specifications
Specifications
| Accession Number | Q61006.1 |
| Conjugate | Unconjugated |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 4593 |
| Quantity | 50 μg |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 2 weeks, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Synonym | EC 2.7.10, EC 2.7.10.1, MGC126323, MGC126324, muscle, skeletal, receptor tyrosine kinase, MuSK, skeletal receptor tyrosine-protein kinase |
| Target | Mouse |
| Buffer | Lyophilized from a 0.2 μm filtered solution in PBS. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.